Merck
Quinidine inhibition of debrisoquine S(+)-4- and 7-hydroxylations in Chinese of different CYP2D6 genotypes.
Pharmacogenetics (1993-04-01)
L Bertilsson, C O Meese, Q Y Yue, M L Dahl, M Ingelman-Sundberg, I Johansson, J Säwe, M Eichelbaum
ABSTRACT
Pronounced differences in the CYP2D6 gene between Chinese and Caucasians have previously been described. There was a low frequency of detrimental mutations in the Chinese CYP2D6 gene causing the poor metabolizer (PM) phenotype. In contrast to Caucasians where the Xba I 44 kb allele is almost always associated with the PM phenotype, Chinese with the 44/44 kb RFLP pattern are extensive metabolizers (EM). In order to evaluate whether the debrisoquine hydroxylation seen in subjects with this haplotype is catalysed by a functionally similar enzyme to CYP2D6 or is catalysed by another type of P450 isozyme, product selectivity of the 4-hydroxylation was studied in 27 Chinese. The inhibition of CYP2D6 by quinidine was also investigated. In the 26 Chinese EM the S(+)-4-hydroxy enantiomer was found to be the major urinary metabolite of debrisoquine with an enantiomeric excess of 96.8-100%, which is similar to that in Caucasians. A correlation between the amount of S(+)-4-hydroxy and the minor 7-hydroxy metabolites excreted in urine (r = 0.72; p < 0.001) was seen. The amount of these two metabolites excreted was less in Chinese EM of debrisoquine with the 44/44 kb RFLP pattern, than in those with the wild type 29/29 kb pattern (p < 0.01). The stereoselectivity was very high in both groups. All Chinese homozygous for the 44 kb fragment (n = 5) were transformed to apparent PM after a single 100 mg dose of quinidine similarly to five Caucasian EM. Both the S(+)-4- and 7-hydroxylations of debrisoquine were inhibited by quinidine in both populations. This study shows that the cytochrome P450 catalysing the 4- and 7-hydroxylations of debrisoquine in Chinese EM has the same properties (product stereoselectivity and inhibition by quinidine) as the CYP2D6 in Caucasian EM.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
(±)-4-Hydroxydebrisoquin sulfate