Merck
  • Adrenic acid delta 4 desaturation and fatty acid composition in liver microsomes of spontaneously diabetic Wistar BB rats.
Adrenic acid delta 4 desaturation and fatty acid composition in liver microsomes of spontaneously diabetic Wistar BB rats.
Prostaglandins, leukotrienes, and essential fatty acids (1994-01-01)
V Mimouni, M Narce, Y S Huang, D F Horrobin, J P Poisson
ABSTRACT
We examined the delta 4 (n-6) desaturation and the fatty acid composition of liver microsomes in the insulin-dependent spontaneously diabetic Wistar Bio-Breeding (BB) rat. The desaturation of adrenic acid to n-6 docosapentaenoic acid was decreased in the normo- and hyperglycemic diabetic rats. Insulin treatment with 1.0 IU. 100 g body weight-1 twice a day for 2 days restored the reduced activity during the hypoglycemic period. The pattern of responses was similar to that of linoleic acid delta 6 and dihomo-gamma-linolenic acid delta 5 desaturases, with a non-parallel relationship between the desaturation system and the glycemia. The microsomal fatty acid composition of BB rat liver reflected only partially to the delta 4 desaturation at different states of glycemia. Factors other than impaired desaturation system are involved in the fatty acid metabolism of spontaneously diabetic rats.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
cis-7,10,13,16-Docosatetraenoic acid, ≥98% (GC)