Merck
  • Studies on antiulcer agents. IV. Antiulcer effects of 2-benzylthio5,6,7,8-tetrahydro-4(3H)-quinazolinones and related compounds.
Studies on antiulcer agents. IV. Antiulcer effects of 2-benzylthio5,6,7,8-tetrahydro-4(3H)-quinazolinones and related compounds.
Chemical & pharmaceutical bulletin (1995-11-01)
K Terashima, H Shimamura, A Kawase, Y Tanaka, T Tanimura, T Kamisaki, Y Ishizuka, M Sato
ABSTRACT
With a view to finding more effective antiulcer agents, a series of 2-benzylthio-5,6,7,8-tetrahydro-4(3H)-quinazolinones and related compounds were synthesized and evaluated in a histamine-stimulated gastric secretion model. The sodium salt of the 2-(dimethylamino)benzylthio derivative (8) showed gastric mucosal protection and gastric antisecretion activities, and was also effective against experimental gastric and duodenal ulcers induced by some ulcerogenic agents. Based on a comparison of the antiulcer properties of 8 with those of the lead compounds (1 and 2) and cimetidine, it appears that, for improvement of antiulcer activity, the reduction of gastric acidity is a more important factor than the reduction of gastric volume output or gastric total acid output.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Mepirizole, analytical standard