Merck
  • Initial employment of di-2-pyridyl ketone as a route to nickel(II)/lanthanide(III) clusters: triangular Ni(2)Ln complexes.
Initial employment of di-2-pyridyl ketone as a route to nickel(II)/lanthanide(III) clusters: triangular Ni(2)Ln complexes.
Dalton transactions (Cambridge, England : 2003) (2010-08-04)
Constantinos G Efthymiou, Anastasia N Georgopoulou, Constantina Papatriantafyllopoulou, Aris Terzis, Catherine P Raptopoulou, Albert Escuer, Spyros P Perlepes
ABSTRACT
Using monoanionic forms of di-2-pyridyl ketone as the only primary ligand, triangular Ni(2)M (M = lanthanide, Y) complexes with interesting magnetic properties have been synthesized.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Di(2-pyridyl) ketone, 99%