Merck
  • Philornis sp. bot fly larvae in free living scarlet macaw nestlings and a new technique for their extraction.
Philornis sp. bot fly larvae in free living scarlet macaw nestlings and a new technique for their extraction.
Veterinary parasitology (2013-02-07)
George Olah, Gabriela Vigo, Lizzie Ortiz, Lajos Rozsa, Donald J Brightsmith
ABSTRACT
Bot fly larvae (Philornis genus) are obligate subcutaneous blood-feeding parasites of Neotropical birds including psittacines. We analyze twelve years of data on scarlet macaw (Ara macao) nestlings in natural and artificial nests in the lowland forests of southeastern Peru and report prevalence and intensity of Philornis parasitism. Bot fly prevalence was 28.9% while mean intensity was 5.0 larvae per infected chick. Prevalence in natural nests (11%, N=90 nestlings) was lower than in wooden nest-boxes (39%, N=57) and PVC boxes (39%, N=109). We describe a new technique of removing Philornis larvae using a reverse syringe design snake bite extractor. We compare this new technique to two other methods for removing bots from macaw chicks and find the new method the most suitable.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Sulfanilamide, ≥98%
Supelco
Sulfanilamide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Supelco
Propoxur, PESTANAL®, analytical standard
Supelco
Coumaphos, PESTANAL®, analytical standard
Supelco
Sulfanilamide, VETRANAL®, analytical standard