Merck
Angiotensin converting enzyme (ACE) gene expression in experimentally induced liver cirrhosis in rats.
Pakistan journal of pharmaceutical sciences (2013-09-17)
Syed Muhammad Shahid, Syeda Nuzhat Fatima, Tabassum Mahboob
ABSTRACT
Angiotensin converting enzyme (ACE) is a key player of Renin Angiotensin System (RAS), involved in conversion of active product, angiotensin-II. Alterations in RAS have been implicated in the pathophysiology of various diseases involving heart, kidney, lung and liver. This study is designed to investigate the association of ACE gene expression in induction of liver cirrhosis in rats. Total 12 male albino Wistar rats were selected and divided in two groups. Control group received 0.9% NaCl, where as Test group received thioacidamide (TAA), dissolved in 0.9%NaCl, injected intraperitoneally at a dosage of 200mg/Kg of body weight, twice a week for 12 weeks. The rats were decapitated and blood sample was collected at the end of experimental period and used for liver functions, enzyme activity, antioxidant enzymes and lipid peroxidation estimations. Genomic DNA was isolated from excised tissue determine the ACE genotypes using specific primers. The ACE gene expression in liver tissue was assessed using the quantitative RT-PCR method. The activity of ALT, total and direct bilirubin, SOD and CAT levels were significantly high (p<0.05) and level of MDA was significantly low (p<0.05) in TAA treated rats as compared to control rats. The ACE gene expression after 12 weeks TAA treatment in cirrhotic rats was significantly increased (p<0.05) in comparison to controls. This study describes the importance of RAS in the development of hepatic fibrosis and the benefits of modulation of this system ACE gene expression. The finding of major up-regulation of ACE in the experimental rat liver provides further insight into the complexities of the RAS and its regulation in liver injury. The development of specific modulators of ACE activity and function, in future, will help determine the role of ACE and its genetic variants in the pathophysiology of liver disease.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Bilirubina, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Thioacetamide, ACS reagent, ≥99.0%
Sigma-Aldrich
Thioacetamide, reagent grade, 98%
Sigma-Aldrich
Bilirubina, purum, ≥95.0% (UV)