Merck
  • Sulfur-containing cytotoxic curvularin macrolides from Penicillium sumatrense MA-92, a fungus obtained from the rhizosphere of the mangrove Lumnitzera racemosa.
Sulfur-containing cytotoxic curvularin macrolides from Penicillium sumatrense MA-92, a fungus obtained from the rhizosphere of the mangrove Lumnitzera racemosa.
Journal of natural products (2013-11-08)
Ling-Hong Meng, Xiao-Ming Li, Cui-Ting Lv, Chun-Shun Li, Gang-Ming Xu, Cai-Guo Huang, Bin-Gui Wang
ABSTRACT
Sumalarins A-C (1-3), the new and rare examples of sulfur-containing curvularin derivatives, along with three known analogues (4-6), were isolated and identified from the cytotoxic extract of Penicillium sumatrense MA-92, a fungus obtained from the rhizosphere of the mangrove Lumnitzera racemosa . Their structures were established by detailed interpretation of NMR and MS data, and compound 1 was confirmed by X-ray crystallographic analysis. Compounds 1-3 and 5 showed potent cytotoxicity against some of the tested tumor cell lines. Sulfur substitution at C-11 or a double bond at C-10 significantly increased the cytotoxic activities of the curvularin analogues.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis
Sigma-Aldrich
sulfur powder, 99.998% trace metals basis
Sigma-Aldrich
Zearalenone, fungal mycotoxin
Supelco
Zearalenone solution, 100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Supelco
Sulfur, PESTANAL®, analytical standard
Sigma-Aldrich
Curvularin, from Penicillium citrinum, ≥98% (HPLC)