Merck
  • Pentaketide melanin biosynthesis in Aspergillus fumigatus requires chain-length shortening of a heptaketide precursor.
Pentaketide melanin biosynthesis in Aspergillus fumigatus requires chain-length shortening of a heptaketide precursor.
The Journal of biological chemistry (2001-05-15)
H F Tsai, I Fujii, A Watanabe, M H Wheeler, Y C Chang, Y Yasuoka, Y Ebizuka, K J Kwon-Chung
ABSTRACT
Chain lengths and cyclization patterns of microbial polyketides are generally determined by polyketide synthases alone. Fungal polyketide melanins are often derived from a pentaketide 1,8-dihydroxynaphthalene, and pentaketide synthases are used for synthesis of the upstream pentaketide precursor, 1,3,6,8-tetrahydroxynaphthalene (1,3,6,8-THN). However, Aspergillus fumigatus, a human fungal pathogen, uses a heptaketide synthase (Alb1p) to synthesize its conidial pigment through a pentaketide pathway similar to that which produces 1,8-dihydroxynaphthalene-melanin. In this study we demonstrate that a novel protein, Ayg1p, is involved in the formation of 1,3,6,8-THN by chain-length shortening of a heptaketide precursor in A. fumigatus. Deletion of the ayg1 gene prevented the accumulation of 1,3,6,8-THN suggesting the involvement of ayg1 in 1,3,6,8-THN production. Genetic analyses of double-gene deletants suggested that Ayg1p catalyzes a novel biosynthetic step downstream of Alb1p and upstream of Arp2p (1,3,6,8-THN reductase). Further genetic and biochemical analyses of the reconstituted strains carrying alb1, ayg1, or alb1 + ayg1 indicated that Ayg1p is essential for synthesis of 1,3,6,8-THN in addition to Alb1p. Cell-free enzyme assays, using the crude Ayg1p protein extract, revealed that Ayg1p enzymatically shortened the heptaketide product of Alb1p to 1,3,6,8-THN. Thus, the protein Ayg1p facilitates the participation of a heptaketide synthase in a pentaketide pathway via a novel polyketide-shortening mechanism in A. fumigatus.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
1,8-Dihydroxynaphthalene, 95%
Prezzi e disponibilità al momento non sono disponibili