Merck
  • Effects of cafedrine/theodrenaline, etilefrine and ephedrine on uterine blood flow during epidural-induced hypotension in pregnant sheep.
Effects of cafedrine/theodrenaline, etilefrine and ephedrine on uterine blood flow during epidural-induced hypotension in pregnant sheep.
Fetal diagnosis and therapy (2005-08-23)
D Strümper, W Gogarten, M E Durieux, K Hartleb, H Van Aken, M A E Marcus
ABSTRACT
Maternal hypotension is a major concern in obstetric anesthesia, and concerns have been raised about standard vasopressor therapy with ephedrine. Therefore, we evaluated the maternal and fetal hemodynamic effects of two potential alternatives to ephedrine. Hypotension was induced by epidural administration of lidocaine in 6 chronically instrumented pregnant ewes (at 118-122 days of gestation, term 145 days). Three treatments were studied: 25 mg ephedrine, 5 mg etilefrine and 100 mg cafedrine/5 mg theodrenaline (C/T) intravenously. Mean fetal and maternal blood pressure and heart rate, uterine blood flow, as well as fetal and maternal arterial blood gases were recorded for 60 min. All three vasopressors increased maternal blood pressure, accompanied by a significant increase in uterine blood flow. C/T caused marked maternal tachycardia, whereas ephedrine decreased maternal heart rate. Maternal and fetal blood gases did not change during any of the three treatment regimens. All three vasopressors restored maternal blood pressure and uterine blood flow after epidurally induced maternal hypotension. However, restoration of uterine perfusion was delayed and less pronounced with C/T.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Etilefrine hydrochloride, European Pharmacopoeia (EP) Reference Standard