Merck
  • Evaluation of oral administration of firocoxib for the management of musculoskeletal pain and lameness associated with osteoarthritis in horses.
Evaluation of oral administration of firocoxib for the management of musculoskeletal pain and lameness associated with osteoarthritis in horses.
American journal of veterinary research (2012-04-27)
James A Orsini, William G Ryan, Douglas S Carithers, Raymond C Boston
ABSTRACT
To generate data on the effects of firocoxib administration to horses with osteoarthritis. Client-owned horses with signs of lameness and joint pain associated with osteoarthritis. Firocoxib was administered as an oral paste (0.1 mg/kg, q 24 h) for 14 days. Assessments were performed on day 0 (baseline) and days 7 and 14. 390 of 429 horses from 80 sites in 25 states met the criteria for analysis. Quarter Horse and Thoroughbred were the 2 most commonly represented breeds, comprising half of the study population. Signs of musculoskeletal pain or lameness attributed to osteoarthritis were diagnosed in a single joint in 197 (197/390 [50.5%]) horses and in multiple joints in 193 (193/390 [49.5%]) horses. In those with involvement of a single joint, the tarsus was the most frequently affected joint (79/197 [40.1 %]). Among the 390 horses with complete lameness data, improvement was reported in approximately 80% by day 14. Investigators rated 307 (78.7%) horses as improved, whereas owners or handlers rated 316 (81.0%) horses as improved at the termination of the study. Horses treated with firocoxib paste had significant improvement in lameness scores from baseline values. Improvement was most rapid within the first 7 days after starting treatment and continued, albeit at a slower rate, through treatment day 14. Firocoxib significantly improved lameness scores throughout the 14-day period with few adverse effects. Firocoxib can be a safe cyclooxygenase-2-specific NSAID for the treatment of musculoskeletal pain and lameness associated with osteoarthritis.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Firocoxib, VETRANAL®, analytical standard