Merck
Effect of yohimbine on detomidine induced changes in behavior, cardiac and blood parameters in the horse.
Veterinary anaesthesia and analgesia (2012-10-06)
Heather K DiMaio Knych, Vanessa Covarrubias, Eugene P Steffey
ABSTRACT
To describe selected pharmacodynamic effects of detomidine and yohimbine when administered alone and in sequence. Randomized crossover design. Nine healthy adult horses aged 9 ± 4 years and weighing 561 ± 56 kg. Three dose regimens were employed in the current study. 1) 0.03 mg kg(-1) detomidine IV, 2) 0.2 mg kg(-1) yohimbine IV and 3) 0.03 mg kg(-1) detomidine IV followed 15 minutes later by 0.2 mg kg(-1) yohimbine IV. Each horse received all three treatments with a minimum of 1 week between treatments. Blood samples were obtained and plasma analyzed for detomidine and yohimbine concentrations by liquid chromatography-mass spectrometry. Behavioral effects, heart rate and rhythm, glucose, packed cell volume and plasma proteins were monitored. Yohimbine rapidly reversed the sedative effects of detomidine in the horse. Additionally, yohimbine effectively returned heart rate and the percent of atrio-ventricular conduction disturbances to pre-detomidine values when administered 15 minutes post-detomidine administration. Plasma glucose was significantly increased following detomidine administration. The detomidine induced hyperglycemia was effectively reduced by yohimbine administration. Effects on packed cell volume and plasma proteins were variable. Intravenous administration of yohimbine effectively reversed detomidine induced sedation, bradycardia, atrio-ventricular heart block and hyperglycemia.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Detomidine hydrochloride, European Pharmacopoeia (EP) Reference Standard