- Moving average improves the signal-to-noise ratio of kinetic studies in magnetic resonance spectroscopy.
Moving average improves the signal-to-noise ratio of kinetic studies in magnetic resonance spectroscopy.
Magnetic resonance in medicine (1990-05-01)
F Seguin, F Berthiau, A Le Pape, J P Grivet
PMID2345519
ABSTRACT
Moving average can be used to improve the signal-to-noise ratio of NMR kinetic studies. The method was tested on simulated spectra with time-dependent intensities or peak positions. It was then applied to a series of in vivo spectra, showing the pharmacological effect of heptaminol on the intracellular pH of perfused hearts.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Heptaminol hydrochloride, European Pharmacopoeia (EP) Reference Standard