- Postanoxic myoclonus: two case presentations and review of medical management.
Postanoxic myoclonus: two case presentations and review of medical management.
Archives of physical medicine and rehabilitation (2013-09-26)
Adrian Budhram, David Lipson, Shanker Nesathurai, David Harvey, Michel P Rathbone
PMID24060492
ABSTRACT
Postanoxic myoclonus is a rare manifestation after an anoxic event, with fewer than 150 cases reported in the literature. The condition is characterized by myoclonic jerks, which are worse on action than at rest, and postural lapses, ataxia, and dysarthria. The disability caused by postanoxic myoclonus can be profound, and treatment in the rehabilitation setting is exceptionally challenging. We present 2 patients who suffered from postanoxic myoclonus after an anoxic event, both of whom were successfully treated with a combination of levetiracetam, valproic acid, and clonazepam. These cases act as a framework for discussing the management of postanoxic myoclonus in the clinical setting.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto