Merck
A motive for the use of a posterior approach in shoulder arthography: ventral leakage of contrast medium.
Acta radiologica (Stockholm, Sweden : 1987) (2013-08-10)
Miraude E A P M Adriaensen, Oliver T Ertl, Peter-Jan van Laar, Maarten Nix, Filip M Vanhoenacker
ABSTRACT
Some orthopedic surgeons request a posterior approach for shoulder magnetic resonance (MR) arthrography, especially in patients with anterior shoulder instability, to avoid interpretive difficulties in differentiating anterior extraarticular contrast injection when using an anterior approach from ventral leakage of contrast. To determine the occurrence of ventral leakage of contrast in shoulder MR arthrography when using a posterior approach. Retrospectively, we included 73 consecutive patients who underwent shoulder MR arthrography (1.0 Tesla) using the posterior approach. Three unsuccessful procedures were excluded. Ventral leakage of contrast, defined as contrast seen around the musculus subscapularis without distention of the posterior capsule, was recorded. Descriptive statistics were used. Seventy shoulders were included. Forty-one left shoulders were involved (59%). Mean age of patients was 49 years (range, 17-76 years). Thirty-five patients were women (50%). Ventral leakage of contrast was seen in 12 shoulders (17%). As ventral leakage of contrast was seen in a substantial number of cases when using a posterior approach in shoulder MR arthrography, the use of a posterior approach is advised to avoid misinterpretation of ventral contrast leakage with accidental extra articular contrast injection, and to increase confidence in the final radiological diagnosis.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
N-metil-D-glucamina, 99.0-100.5% (titration)
Sigma-Aldrich
Histodenz, nonionic density gradient medium
Sigma-Aldrich
N-metil-D-glucamina, ReagentPlus®, ≥99.0% (T)
Sigma-Aldrich
N-metil-D-glucamina, 99.0-100.5% dry basis, meets USP testing specifications
Supelco
Iohexol, analytical standard
Supelco
N-metil-D-glucamina, Pharmaceutical Secondary Standard; Certified Reference Material
Iohexol, European Pharmacopoeia (EP) Reference Standard
N-metil-D-glucamina, European Pharmacopoeia (EP) Reference Standard
Iohexol for peak identification, European Pharmacopoeia (EP) Reference Standard