- Iron-catalyzed oxidative direct α-C-H bond functionalization of cyclic ethers: selective C-O bond formation in the presence of a labile aldehyde group.
Iron-catalyzed oxidative direct α-C-H bond functionalization of cyclic ethers: selective C-O bond formation in the presence of a labile aldehyde group.
Organic letters (2014-03-19)
Balaji D Barve, Yang-Chang Wu, Mohamed El-Shazly, Michal Korinek, Yuan-Bin Cheng, Jeh-Jeng Wang, Fang-Rong Chang
PMID24635116
ABSTRACT
Iron catalyzed oxidative coupling of salicylaldehydes with cyclic ethers proceeded through the direct α-C-H functionalization of ethers, forming the corresponding acetals in moderate to excellent yields. This is the first example of iron catalyzed selective C-O bond formation in the presence of a sensitive aldehyde moiety.