Merck
Leuprorelin acetate granulomas: case reports and review of the literature.
The British journal of dermatology (2005-05-13)
K Yasukawa, D Sawamura, H Sugawara, N Kato
ABSTRACT
Leuprorelin acetate, an agonist of gonadotropin-releasing hormone (GnRH), is indicated in the treatment of prostate cancer. Recently, depot formulations of leuprorelin acetate have been widely used. We report three patients who showed a granulomatous reaction after treatment using a leuprorelin acetate 3-month depot formulation. These patients presented with 5-6-cm subcutaneous nodules at injection sites, which developed after the depot type was changed from a 1-month to a 3-month formulation. Skin biopsy showed epithelioid cells and foreign body giant cells containing round, translucent microspheres which formed sarcoidal granulomas. Changing to other GnRH agonists resulted in no further problems. We have reviewed the previous reports of leuprorelin acetate-induced granuloma formation. The formation of such granulomas may be related to the polymers that allow slow release after injection, or leuprorelin acetate itself may be responsible. The depot injection methods using leuprorelin also seem to have a causal effect in granuloma formation. Dermatologists need to know that depot leuprorelin acetate may cause a granulomatous reaction which produces a subcutaneous nodule that might be misdiagnosed as a malignant tumour.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Leuprorelin, European Pharmacopoeia (EP) Reference Standard