Merck
  • Triptolide: structural modifications, structure-activity relationships, bioactivities, clinical development and mechanisms.
Triptolide: structural modifications, structure-activity relationships, bioactivities, clinical development and mechanisms.
Natural product reports (2012-01-25)
Zhao-Li Zhou, Ya-Xi Yang, Jian Ding, Yuan-Chao Li, Ze-Hong Miao
ABSTRACT
Triptolide, a principal bioactive ingredient of Tripterygium wilfordii Hook F, has attracted extensive exploration due to its unique structure of a diterpenoid triepoxide and multiple biological activities. This review will focus on the structural modifications, structure-activity relationships, pharmacology, and clinical development of triptolide in the last forty years.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Triptolide, from Tripterygium wilfordii, ≥98% (HPLC), solid