Merck
Radiolytic studies of naphthalene in the presence of water.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life (2012-06-05)
Y Keheyan, I L ten Kate
ABSTRACT
Naphthalene is an interesting candidate to study in the framework of organic delivery to planetary surfaces as well as in the origin of life. Additionally, naphthalene is of environmental interest, because of its chronic and acute effects on living systems, such as humans and animals (e.g. moths). Naphthalene has been well studied in both fields. In this paper we give an overview of radiolytic studies of naphthalene in the presence of both liquid water and water ice. From our review it appears that OH radicals are formed both in liquid water and in interstellar ices and that these radicals play a considerable role in the degradation of naphthalene. However, it also appears that upon irradiation of naphthalene in liquid water, hydrogen peroxide, a species that accelerates naphthalene degradation, is formed. Based on this review we suggest that the role of hydrogen peroxide in interstellar ices should be further investigated.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Naphthalene, 99%
Supelco
Naphthalene, analytical standard
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Supelco
Melting point standard 79.5-81°C, analytical standard
Supelco
Naphthalene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland