Merck
Subacute leucoencephalopathy induced by carmofur, a 5-fluorouracil derivative.
Journal of neurology (1987-08-01)
S Kuzuhara, N Ohkoshi, K Kanemaru, H Hashimoto, T Nakanishi, Y Toyokura
ABSTRACT
Three cases of leucoencephalopathy induced by carmofur (1-hexylcarbamoyl-5-fluorouracil), an antineoplastic derivative of 5-fluorouracil are reported and the literature is reviewed. Initial symptoms were unsteady gait and dementia developing several weeks or months after carmofur had been started. Symptoms increased gradually even after stopping the drug. Severe encephalopathy with confusion, delirium or coma appeared frequently. Symptoms were usually reversible but death occasionally occurred. The EEG showed marked slowing. Computed tomography of the brains of severely intoxicated patients showed marked hypodensity of the entire cerebral white matter. Carmofur must be discontinued immediately if any psychomotor symptoms develop.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Carmofur, ≥98% (HPLC), powder