Merck
Capacitative Ca2+ entry in HL-60 cells: tetrandrine and SK & F 96365 as probes.
Zhongguo yao li xue bao = Acta pharmacologica Sinica (1996-03-01)
Y M Leung, C Y Kwan, T T Loh
ABSTRACT
Agonist-activated Ca2+ entry is important in many biological responses such as secretion and cell growth(1,2). In nonexcitable cells which have no voltage-operated Ca2+ channels (VOCC), agonist-receptor interaction can trigger Ca2+ entry across the plasmalemma via several entry pathways(1-3) (Fig 1): (A) channels which are intrinsic structures of the receptor (receptor-operated channels), (B) channels which are coupled to receptors via a G-protein (G-protein-operated channels), (C) channels which are activated by some second messengers (second-messenger-operated channels), and (D) channels which open upon intracellular nonmitochondrial Ca2+ store depletion (Ca2+ release-activated channels) resulting from inositol 1, 4, 5-trisphosphate-induced Ca2+ release or inhibition of Ca2+ re-uptake (see next section). Ca2+ entry via the 4th type of channel, also known as "capacitative Ca2+ entry" (CCE)[4], has aroused much interest in the past decade because of its intriguing nature as retrograde signalling. In this brief review, we present the evidence for and the possible biochemical processes involved in CCE. We also discuss the use of 2 novel Ca2+ entry blockers: tetrandrine and SK&F 96365. Emphasis will be put on the human leukemic HL-60 cell line, a popular cell system for intracellular Ca2+ homeostasis studies and also a model the signal transduction of which we have been investigating during the past few years.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Tetrandrine, analytical standard, for drug analysis
Tetrandrine, European Pharmacopoeia (EP) Reference Standard