Merck
Pituitary gastrins. Different processing in corticotrophs and melanotrophs.
The Journal of biological chemistry (1981-10-25)
J F Rehfeld, L I Larsson
ABSTRACT
The anterior, intermediate, and neural lobes of porcine pituitaries contain different molecular forms of gastrin. In the anterior lobe, unsulfated large gastrins, component I, and gastrin-34 constitute the main forms. In contrast in the intermediate and neural lobes, gastrin-17, sulfated as well as unsulfated, predominates. Since component I and gastrin-34 are biosynthetic precursors of gastrin-17, the findings indicate that proteolytic processing and amino acid derivatization (sulfation) differs between the lobes. This difference, together with the localization of anterior lobe gastrin in corticotrophs and intermediate lobe gastrin in melanotrophs, emphasizes the close relation and parallelism of the biosynthesis of gastrin and corticotropin peptides.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Gastrin Rabbit Polyclonal Antibody