- Asymmetric total synthesis of (-)-leuconoxine via chiral phosphoric acid catalyzed desymmetrization of a prochiral diester.
Asymmetric total synthesis of (-)-leuconoxine via chiral phosphoric acid catalyzed desymmetrization of a prochiral diester.
Organic letters (2014-12-20)
Kazuhiro Higuchi, Shin Suzuki, Reeko Ueda, Norifumi Oshima, Emiko Kobayashi, Masanori Tayu, Tomomi Kawasaki
PMID25522825
ABSTRACT
The asymmetric total synthesis of (-)-leuconoxine has been achieved. The desymmetrization of a prochiral diester using a chiral phosphoric acid catalyst produced a highly enantioenriched lactam with excellent yield. The ring construction featuring an intramolecular N-acyliminium cyclization and the one-step pyrrolidone formation using Bestmann's ylide was successfully accomplished.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.