Merck
  • Differential effects of a common splice site polymorphism on the generation of OAS1 variants in human bronchial epithelial cells.
Differential effects of a common splice site polymorphism on the generation of OAS1 variants in human bronchial epithelial cells.
Human immunology (2012-12-12)
Satoshi Noguchi, Emi Hamano, Ikumi Matsushita, Minako Hijikata, Hideyuki Ito, Takahide Nagase, Naoto Keicho
ABSTRACT
The 2',5'-oligoadenylate synthetase 1 (OAS1) is one of the major interferon-inducible proteins and a critical component of the host defense system against viral infection. A single nucleotide polymorphism (SNP), rs10774671, presumably responsible for alternate splicing of this gene, has frequently been associated with a variety of viral diseases, including emerging respiratory infections. We investigated the SNP-dependent expression of OAS1 variants in primary cultured human bronchial epithelial cells. Total RNA was subjected to real-time RT-PCR with specific primer sets designed to amplify each transcript variant. We found that the p46 transcript was mainly expressed in cells with the GG genotype, whereas the p42 transcript was highly expressed, and the p44a (alternate exon in intron 5), p48, and p52 transcripts were expressed to a lesser extent, in cells with the AA genotype. Immunoblot analysis revealed that the p46 isoform and a smaller amount of the p42 isoform were present in cells with the GG genotype, whereas only the p42 isoform was clearly observed in cells with the AA genotype. Cellular DNA fragmentation induced by neutrophil elastase was more preferentially found in cells with the AA genotype. Thus, our findings provide insights into the potential role of OAS1 polymorphisms in respiratory infection.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-OAS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution