Merck
Identification and characterization of a novel class of atypical dopamine receptor agonists.
Pharmaceutical research (2012-05-02)
E V Kuzhikandathil, S Kortagere
ABSTRACT
The D3 dopamine receptor exhibits tolerance and slow response termination (SRT) properties that are not exhibited by the closely-related D2 dopamine receptor. We previously demonstrated that the induction of tolerance elicits a unique conformational change in the D3 receptor. Here we tested the hypothesis that the tolerance and SRT properties of the D3 receptor are ligand-dependent. We used pharmacophore modeling and in silico screening approaches coupled with electrophysiological and biochemical methods to identify and functionally characterize the novel dopamine receptor agonists. We identified cis-8-OH-PBZI (PBZI), FAUC73 and an additional novel compound, ES609, which although they are full D3 receptor agonists, do not induce the tolerance and SRT properties of the D3 receptor. In addition, PBZI has full intrinsic activity at D2L, is a partial agonist at D2S and exhibits functional selectivity at D4.2 dopamine receptors. ES609 is a partial agonist at D2S, D2L and D4.2 receptors, and exhibits functional selectivity at D2L and D4.2 dopamine receptors. We have discovered a novel class of atypical dopamine receptor agonists that include three structurally dissimilar compounds. These new agonists will help determine the physiological and pathophysiological relevance of D3 receptor tolerance and SRT properties.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
cis-8-Hydroxy-3-(n-propyl)-1,2,3a,4,5,9b-hexahydro-1H-benz[e]indole hydrobromide, ≥98% (HPLC), solid