Merck
  • Lymphocytic colitis is associated with increased pro-inflammatory cytokine profile and up regulation of prostaglandin receptor EP4.
Lymphocytic colitis is associated with increased pro-inflammatory cytokine profile and up regulation of prostaglandin receptor EP4.
PloS one (2013-04-25)
Indranil Dey, Paul L Beck, Kris Chadee
ABSTRACT
Microscopic colitis (MC) is comprised of two entities, lymphocytic (LC) and collagenous colitis. Up to 20% of patients with chronic diarrhea that have a normal appearing colonoscopy will be diagnosed with MC. Since MC was first described less than 40 years ago, little is known about the mechanisms involved in disease pathogenesis. Nonsteroidal anti-inflammatory drugs are associated with an increased risk of MC and some reports suggest a dysregulation in prostaglandin production. Recent genome wide screens have found an association between prostaglandin receptor EP4 expression and inflammatory bowel disease; however, EP4 expression has never been studied in MC. The aim of this study was to assess colonic mucosal inflammatory cytokine profiles in patients with LC and to assess expression of the prostaglandin receptor EP4. Colonic mucosal biopsies were obtained from patients undergoing colonoscopy for investigation of diarrhea and in those undergoing colon cancer screening. Following histological assessment, expression of cytokines and the prostaglandin receptor EP4 was analyzed using real-time reverse transcriptase-PCR and immunohistochemistry. Patients with LC had markedly increased mRNA expression for TNF-α, IFN-γ and IL-8 compared to normal controls (p<0.001). No significant differences were noted for IL-1β, IL-4, IL-10 or IL-12/23. Interestingly, those with LC had increased EP4 receptor expression, which positively correlated with increased TNF-α expression. This is the first report to demonstrate that LC is associated with increased TNF-α, INF-γ and IL-8 concurrent with a marked up-regulation of EP4. These findings add to our knowledge on the pathogenesis of LC and may give rise to possible new therapeutic and/or diagnostic tools in the management of MC.