Merck
Kinetics of serotonin oxidation by heme-Aβ relevant to Alzheimer's disease.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2014-09-25)
Soumya Mukherjee, Manas Seal, Somdatta Ghosh Dey
ABSTRACT
Serotonin (5-HT) is an essential neurotransmitter for cognitive functions and formation of new memories. A deficit in 5-HT dependent neuronal activity is somewhat specific for Alzheimer's disease. Metal-mediated oxidative degradation of neurotransmitters by Aβ bound to metals has been investigated. Heme-bound Aβ is found to catalyze the oxidative degradation of 5-HT leading to the formation of neurotoxic products dihydroxybitryptamine and tyrptamine-4,5-dione. The catalytic degradation of 5-HT is of first order with respect to both heme-Aβ and H2O2, and the maximum rate of 5-HT oxidation is obtained at physiological pH (pH 7-7.5). pH perturbation of the binding affinity of heme-Aβ complex for 5-HT indicates that the binding of the substrate (5-HT) is not the rate-determining step. Arg5 acts as a second-sphere residue facilitating the O-O bond cleavage, the mutation of which leads to a decrease in the rate of 5-HT oxidation. The pull effect of the Arg5 residue tends to facilitate the generation of the active oxidant, Compound I, below neutral pH, while the ionization of the phenol group of the substrate facilitates the generation of the active substrate above neutral pH. A combination of these two opposing effects results in the highest activity at physiological pH. Apart from the Arg5 residue, the Tyr10 residue is found to play a vital role in the 5-HT oxidation by heme-Aβ complexes.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acido solforico, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Perossido di idrogeno, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Perossido di idrogeno, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acido solforico, 99.999%
Sigma-Aldrich
Perossido di idrogeno, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Emina, from bovine, ≥90%
Sigma-Aldrich
Emina, BioXtra, from Porcine, ≥96.0% (HPLC)
Millipore
Perossido di idrogeno, 3%, suitable for microbiology
Supelco
Perossido di idrogeno, ≥30%, for trace analysis
Sigma-Aldrich
Soluzione tampone HEPES, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Perossido di idrogeno, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Supelco
Acido solforico, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Perossido di idrogeno, 34.5-36.5%
Supelco
Perossido di idrogeno, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Supelco
Acido solforico, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard