Merck
  • Biomarkers probed in saliva by surface plasmon resonance imaging coupled to matrix-assisted laser desorption/ionization mass spectrometry in array format.
Biomarkers probed in saliva by surface plasmon resonance imaging coupled to matrix-assisted laser desorption/ionization mass spectrometry in array format.
Analytical and bioanalytical chemistry (2014-12-20)
Johana Musso, William Buchmann, Florence Gonnet, Nathalie Jarroux, Sophie Bellon, Chiraz Frydman, Didier-Luc Brunet, Regis Daniel
ABSTRACT
Detection of protein biomarkers is of major interest in proteomics. This work reports the analysis of protein biomarkers directly from a biological fluid, human saliva, by surface plasmon resonance imaging coupled to mass spectrometry (SPRi-MS), using a functionalized biochip in an array format enabling multiplex SPR-MS analysis. The SPR biochip presented a gold surface functionalized by a self-assembled monolayer of short poly(ethylene oxide) chains carrying an N-hydroxysuccinimide end-group for the immobilization of antibodies. The experiments were accomplished without any sample pre-purification or spiking with the targeted biomarkers. SPRi monitoring of the interactions, immune capture from the biochip surface, and finally on-chip matrix-assisted laser desorption/ionization-MS structural identification of two protein biomarkers, salivary α-amylase and lysozyme, were successively achieved directly from saliva at the femtomole level. For lysozyme, the on-chip MS identification was completed by a proteomic analysis based on an on-chip proteolysis procedure and a peptide mass fingerprint.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Glicina, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ammonio acetato, ACS reagent, ≥97%
Sigma-Aldrich
Glicina, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Ammonio acetato, ≥99.99% trace metals basis
Supelco
Ammonio acetato, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
L-lisina, ≥98% (TLC)
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonio acetato, Molecular Biology, ≥98%
Sigma-Aldrich
Glicina, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glicina, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-lisina, crystallized, ≥98.0% (NT)
Sigma-Aldrich
Ammonio acetato, 99.999% trace metals basis
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
SAFC
Glicina
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Ammonio acetato, reagent grade, ≥98%
Supelco
Glicina, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonio acetato, BioXtra, ≥98%
Sigma-Aldrich
Glicina, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glicina, ACS reagent, ≥98.5%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
USP
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
L-lisina, analytical standard
Sigma-Aldrich
Ammonio acetato, BioUltra, ≥99.0%
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Glicina, 99%, FCC