Merck
  • Mitotane enhances doxorubicin cytotoxic activity by inhibiting P-gp in human adrenocortical carcinoma cells.
Mitotane enhances doxorubicin cytotoxic activity by inhibiting P-gp in human adrenocortical carcinoma cells.
Endocrine (2014-08-07)
Teresa Gagliano, Erica Gentilin, Katiuscia Benfini, Carmelina Di Pasquale, Martina Tassinari, Simona Falletta, Carlo Feo, Federico Tagliati, Ettore Degli Uberti, Maria Chiara Zatelli
ABSTRACT
Mitotane is currently employed as adjuvant therapy as well as in the medical treatment of adrenocortical carcinoma (ACC), alone or in combination with chemotherapeutic agents. It was previously demonstrated that mitotane potentiates chemotherapeutic drugs cytotoxicity in cancer cells displaying chemoresistance due to P-glycoprotein (P-gp), an efflux pump involved in cancer multidrug resistance. The majority of ACC expresses high levels of P-gp and is highly chemoresistent. The aim of our study was to explore in vitro whether mitotane, at concentrations lower than those currently reached in vivo, may sensitize ACC cells to the cytotoxic effects of doxorubicin and whether this effect is due to a direct action on P-gp. NCI-H295 and SW13 cell lines as well as 4 adrenocortical neoplasia primary cultures were treated with mitotane and doxorubicin, and cell viability was measured by MTT assay. P-gp activity was measured by calcein and P-gp-Glo assays. P-gp expression was evaluated by Western blot. We found that very low mitotane concentrations sensitize ACC cells to the cytotoxic effects of doxorubicin, depending on P-gp expression. In addition, mitotane directly inhibits P-gp detoxifying function, allowing doxorubicin cytotoxic activity. These data provide the basis for the greater efficacy of combination therapy (mitotane plus chemotherapeutic drugs) on ACC patients. Shedding light on mitotane mechanisms of action could result in an improved design of drug therapy for patients with ACC.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
USP
Verapamil hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acido bicinconinico, ≥98% (HPLC)
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard