Merck
  • Phosphorylation of a threonine unique to the short C-terminal isoform of betaII-spectrin links regulation of alpha-beta spectrin interaction to neuritogenesis.
Phosphorylation of a threonine unique to the short C-terminal isoform of betaII-spectrin links regulation of alpha-beta spectrin interaction to neuritogenesis.
The Journal of biological chemistry (2006-11-08)
Paola A Bignone, Mikayala D A King, Jennifer C Pinder, Anthony J Baines
ABSTRACT
Spectrin tetramers are cytoskeletal proteins required in the formation of complex animal tissues. Mammalian alphaII- and betaII-spectrin subunits form dimers that associate head to head with high affinity to form tetramers, but it is not known if this interaction is regulated. We show here that the short C-terminal splice variant of betaII-spectrin (betaIISigma2) is a substrate for phosphorylation. In vitro, protein kinase CK2 phosphorylates Ser-2110 and Thr-2159; protein kinase A phosphorylates Thr-2159. Antiphospho-Thr-2159 peptide antibody detected phosphorylated betaIISigma2 in Cos-1 cells. Immunoreactivity was increased in Cos-1 cells by treatment with forskolin, indicating that phosphorylation is promoted by elevated cAMP. The effect of forskolin was counteracted by the cAMP-dependent kinase inhibitor, H89. In vitro, protein kinase A phosphorylation of an active fragment of betaIISigma2 greatly reduced its interaction with alphaII-spectrin at the tetramerization site. Mutation of Thr-2159 to alanine eliminated inhibition by phosphorylation. Among the processes that require spectrin in mammals is the formation of neurites (incipient nerve axons). We tested the relationship of spectrin phosphorylation to neuritogenesis by transfecting the neuronal cell line, PC12, with enhanced green fluorescent protein-coupled fragments of betaIISigma2-spectrin predicted to act as inhibitors of spectrin tetramer formation. Both wild-type and T2159E mutant fragments allowed normal neuritogenesis in PC12 cells in response to nerve growth factor. The mutant T2159A inhibited neuritogenesis. Because the T2159A mutant represents a high affinity inhibitor of tetramer formation, we conclude that tetramers are requisite for neuritogenesis. Furthermore, because both the T2159E mutant and the wild-type allow neuritogenesis, we conclude that the short C-terminal betaII-spectrin is phosphorylated during this process.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Casein Kinase II, Human, Recombinant, E. coli, Casein Kinase II, Human, Recombinant, E. coli Recombinant, is expressed in E. coli. A cyclic nucleotide-independent Ser/Thr protein kinase composed of catalytic α-subunits and regulatory β-subunits.