Merck
Two new proteases in the MHC class I processing pathway.
Nature immunology (2001-03-23)
L Stoltze, M Schirle, G Schwarz, C Schröter, M W Thompson, L B Hersh, H Kalbacher, S Stevanovic, H G Rammensee, H Schild
ABSTRACT
The proteasome generates exact major histocompatibility complex (MHC) class I ligands as well as NH2-terminal-extended precursor peptides. The proteases responsible for the final NH2-terminal trimming of the precursor peptides had, until now, not been determined. By using specific selective criteria we purified two cytosolic proteolytic activities, puromycin-sensitive aminopeptidase and bleomycin hydrolase. These proteases could remove NH2-terminal amino acids from the vesicular stomatitis virus nucleoprotein cytotoxic T cell epitope 52-59 (RGYVYQGL) resulting, in combination with proteasomes, in the generation of the correct epitope. Our data provide evidence for the existence of redundant systems acting downstream of the proteasome in the antigen-processing pathway for MHC class I molecules.