Merck
  • Relationship between the intracellular effects of camptothecin and the inhibition of DNA topoisomerase I in cultured L1210 cells.
Relationship between the intracellular effects of camptothecin and the inhibition of DNA topoisomerase I in cultured L1210 cells.
Cancer research (1987-04-01)
M R Mattern, S M Mong, H F Bartus, C K Mirabelli, S T Crooke, R K Johnson
ABSTRACT
Results of filter elution assays of lesions produced in the DNA of cultured L1210 cells by the antineoplastic alkaloid camptothecin support the notion that topoisomerase I is an intracellular target of this drug. One to 10 microM camptothecin induced DNA single-strand, but not double-strand, breaks when incubated with intact cells or with their isolated nuclei. Approximately one half of the strand breakage was protein concealed, as judged by filter elution. Camptothecin-induced, protein-concealed DNA strand breaks disappeared rapidly after drug removal. DNA-protein cross-links were generated by camptothecin with frequencies approximately equal to those of protein-concealed DNA strand breaks. It is likely that camptothecin can inhibit topoisomerase I in intact cells in a manner similar to that in which other antineoplastic agents such as amsacrine or teniposide inhibit topoisomerase II. DNA-breaking lesions other than those resulting from trapped topoisomerase I-DNA complexes may also be generated by camptothecin. The yields of DNA strand breaks induced by camptothecin, amsacrine, or teniposide were approximately doubled when cells were incubated for 16 h with 3-aminobenzamide, an inhibitor of poly(ADP ribosylation) of proteins, prior to 1-h exposure to the antineoplastic compounds. 3-Aminobenzamide also enhanced the cytotoxic action of camptothecin, amsacrine, and teniposide. These results suggest that protein-concealed strand breaks can be lethal lesions and that intracellular topoisomerase I and II activity may be regulated coordinately through poly(ADP ribosylation).
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Tetrapropylammonium hydroxide solution, 1.0 M in H2O