Merck
Molecular mechanism of statin-mediated LOX-1 inhibition.
Cell cycle (Georgetown, Tex.) (2015-05-08)
Silvia Biocca, Federico Iacovelli, Sara Matarazzo, Giulia Vindigni, Francesco Oteri, Alessandro Desideri, Mattia Falconi
ABSTRACT
Statins are largely used in clinics in the treatment of patients with cardiovascular diseases for their effect on lowering circulating cholesterol. Lectin-like oxidized low-density lipoprotein (LOX-1), the primary receptor for ox-LDL, plays a central role in the pathogenesis of atherosclerosis and cardiovascular disorders. We have recently shown that chronic exposure of cells to lovastatin disrupts LOX-1 receptor cluster distribution in plasma membranes, leading to a marked loss of LOX-1 function. Here we investigated the molecular mechanism of statin-mediated LOX-1 inhibition and we demonstrate that all tested statins are able to displace the binding of fluorescent ox-LDL to LOX-1 by a direct interaction with LOX-1 receptors in a cell-based binding assay. Molecular docking simulations confirm the interaction and indicate that statins completely fill the hydrophobic tunnel that crosses the C-type lectin-like (CTLD) recognition domain of LOX-1. Classical molecular dynamics simulation technique applied to the LOX-1 CTLD, considered in the entire receptor structure with or without a statin ligand inside the tunnel, indicates that the presence of a ligand largely increases the dimer stability. Electrophoretic separation and western blot confirm that different statins binding stabilize the dimer assembly of LOX-1 receptors in vivo. The simulative and experimental results allow us to propose a CTLD clamp motion, which enables the receptor-substrate coupling. These findings reveal a novel and significant functional effect of statins.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
2-Propanolo, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
2-Propanolo, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanolo, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
2-Propanolo, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, BioReagent, suitable for fluorescence, ≥98.0% (TLC)
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, 97%
Sigma-Aldrich
Sodium trichloroacetate, 97%