Merck
  • One Step Preparation of Reduced Graphene Oxide/Pd-Fe3 O4 @Polypyrrole Composites and Their Application in Catalysis.
One Step Preparation of Reduced Graphene Oxide/Pd-Fe3 O4 @Polypyrrole Composites and Their Application in Catalysis.
Chemistry, an Asian journal (2015-06-26)
Tongjie Yao, Hao Wang, Quan Zuo, Jie Wu, Xiao Zhang, Fang Cui, Tieyu Cui
ABSTRACT
The simple preparation of catalysts with superior catalytic activity and good reusability is highly desirable. Herein, we report a novel strategy to construct reduced graphene oxide (rGO)/Pd-Fe3 O4 @polypyrrole (PPy) catalysts with Pd and Fe3 O4 nanoparticles anchored on a rGO nanosheet surface and wrapped in a PPy shell. The synthesis and assembly of both the Pd and Fe3 O4 nanoparticles, the preparation of the PPy layer, and the reduction of graphene oxide nanosheets were finished in one step. In the system, the PPy layer not only prevented aggregation of Pd and Fe3 O4 nanoparticles, but also generated a synergistic effect with precursor Pd(2+) ions, which led to a high dispersity of as-prepared Pd nanoparticles. Although the procedure was simplified to one step, the catalytic activity and reusability were not sacrificed. In the reduction of 4-nitrophenol, their catalytic performance was better than that in recent reports. Moreover, the catalysts showed good reusability owing to their magnetic properties.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Perossido di idrogeno, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Boroidruro di sodio, powder, ≥98.0%
Sigma-Aldrich
Acido solforico, 99.999%
Sigma-Aldrich
Boroidruro di sodio, ReagentPlus®, 99%
Sigma-Aldrich
Cloruro di palladio(II), ReagentPlus®, 99%
Sigma-Aldrich
Cloruro di palladio(II), ≥99.9%
Sigma-Aldrich
Pyrrole, reagent grade, 98%
Sigma-Aldrich
Boroidruro di sodio, granular, 99.99% trace metals basis
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Boroidruro di sodio, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Perossido di idrogeno, 34.5-36.5%
Sigma-Aldrich
Boroidruro di sodio, granular, 10-40 mesh, 98%
Sigma-Aldrich
Cloruro di palladio(II), anhydrous, 60% Pd basis
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Pyrrole, ≥98%, FCC, FG
Sigma-Aldrich
Boroidruro di sodio, powder
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Palladium(II) chloride solution, 5 wt. % in 10 wt. % HCl
Sigma-Aldrich
Cloruro di palladio(II), 99.995%
Sigma-Aldrich
Boroidruro di sodio, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Cloruro di palladio(II), anhydrous, 59-60% palladium (Pd) basis
Sigma-Aldrich
Palladium(II) chloride ChemBeads