Merck
Relaxation dynamics and exciton energy transfer in the low-temperature phase of MEH-PPV.
The Journal of chemical physics (2015-06-08)
Cristina Consani, Federico Koch, Fabian Panzer, Thomas Unger, Anna Köhler, Tobias Brixner
ABSTRACT
Understanding the effects of aggregation on exciton relaxation and energy transfer is relevant to control photoinduced function in organic electronics and photovoltaics. Here, we explore the photoinduced dynamics in the low-temperature aggregated phase of a conjugated polymer by transient absorption and coherent electronic two-dimensional (2D) spectroscopy. Coherent 2D spectroscopy allows observing couplings among photoexcited states and discriminating band shifts from homogeneous broadening, additionally accessing the ultrafast dynamics at various excitation energies simultaneously with high spectral resolution. By combining the results of the two techniques, we differentiate between an initial exciton relaxation, which is not characterized by significant exciton mobility, and energy transport between different chromophores in the aggregate.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Poly[2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylenevinylene], average Mn 40,000-70,000
Sigma-Aldrich
Poly[2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylenevinylene], average Mn 70,000-100,000