Merck
Pathogenic Role of a Proliferation-Inducing Ligand (APRIL) in Murine IgA Nephropathy.
PloS one (2015-09-09)
Yang Gyun Kim, Montserrat Alvarez, Hitoshi Suzuki, Sachiko Hirose, Shozo Izui, Yasuhiko Tomino, Bertrand Huard, Yusuke Suzuki
ABSTRACT
A proliferation-inducing ligand (APRIL) is a member of the tumor necrosis factor (TNF) superfamily. Despite advances in clinical and genetic studies, the details of the pathological roles of APRIL in IgA nephropathy (IgAN) remain to be fully defined. The present study aimed to further assess the pathological role of APRIL using a mouse model of IgAN. Mice with IgAN designated "grouped ddY" (gddY) were intraperitoneally administered an anti-APRIL monoclonal antibody (anti-APRIL Ab) or control IgG (Control Ab) twice each week for 2 weeks starting during the early stage of IgAN (6-7 weeks of age). Urinary albumin, serum IgA, and glomerular IgA deposition were evaluated. We further assessed the inflammatory responses during treatment by measuring the levels of the chemokine fractalkine (FKN) and its receptor CX3CR1 as well as the level of peripheral blood monocytosis. Anti-APRIL Ab treatment significantly decreased albuminuria and tissue damage combined with decreases in serum IgA levels and deposition of glomerular IgA. In contrast, the abundance of IgA+/B220+ or CD138+/B220+ B cells in the spleen and bone marrow, respectively, was unchanged. Treating gddY mice with anti-April Ab reduced the overexpression of FKN/CX3CR1 in the kidney and the increase in the population of circulating Gr1-/CD115+ monocytes. The size of the population of Gr1-/CD115+ monocytes correlated with renal FKN and urinary albumin levels. Moreover, mice treated with anti-APRIL Ab exhibited reduced progression of IgAN, serum IgA levels, and glomerular IgA deposition as well as an attenuated inflammatory process mediated by FKN-associated activation of monocytes. To the best of our knowledge, this is the first study to implicate the APRIL signal transduction pathway in the pathogenesis of nephrogenic IgA production. Moreover, our findings identify APRIL as a potential target of therapy.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Csf1r