Merck
  • Anti-Androgenic Activity of Nardostachys jatamansi DC and Tribulus terrestris L. and Their Beneficial Effects on Polycystic Ovary Syndrome-Induced Rat Models.
Anti-Androgenic Activity of Nardostachys jatamansi DC and Tribulus terrestris L. and Their Beneficial Effects on Polycystic Ovary Syndrome-Induced Rat Models.
Metabolic syndrome and related disorders (2015-04-29)
Palakkil Mavilavalappil Sandeep, Toine F H Bovee, Krishnan Sreejith
ABSTRACT
Polycystic ovary syndrome (PCOS) is a major hyperandrogenic disorder. Many drugs prescribed specifically to treat PCOS have side effects; however, previous studies suggest that natural therapeutics including botanicals may be less invasive and equally effective for the management of PCOS. In the present study, plants were screened for antiandrogenic activity using the RIKILT yeast Androgen bioAssay (RAA). Selected positive plants were subsequently tested for their efficacy against PCOS induced by estradiol valerate (EV) in rat models. RAA revealed the antiandrogenic property of Nardostachys jatamansi DC (NJ), Tribulus terrestris L. (TT), and Embelia tsjeriam-cottam DC (EJ), whereas Whithania somnifera Dunal (WS), Symplocos racemosa Roxb. (SR), and Helicteres isora L. (HI) exhibited androgenic properties. EJ also exhibited mild androgenic activity and therefore was excluded from further study. EV administration reduced the weight gain and disrupted cyclicity in all rats. NJ and TT extract treatment normalized estrous cyclicity and steroidal hormonal levels and regularized ovarian follicular growth. The in vitro antiandrogenic activity of plant extracts and their positive effects on different parameters of PCOS were proved in vivo.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Flutamide
Sigma-Aldrich
β-Estradiol 17-valerate, ≥98%