Merck
  • Stabilization of Gas Bubbles Released from Water-Soluble Carbohydrates Using Amphiphilic Compounds: Preparation of Formulations and Acoustic Monitoring of Bubble Lifetime.
Stabilization of Gas Bubbles Released from Water-Soluble Carbohydrates Using Amphiphilic Compounds: Preparation of Formulations and Acoustic Monitoring of Bubble Lifetime.
Journal of surfactants and detergents (2011-10-01)
Lars Hoff, Per A Foss, Knut Dyrstad, Jo Klaveness, Pål Rongved
ABSTRACT
The ultrasound contrast agents Echovist(®) and Levovist(®) (Bayer AG, Schering AG, Germany) are based on the release of gas bubbles from milled α-d-galactose. In diagnostic ultrasound, for this class of contrast agents, there is a need for prolonged contrast duration. To investigate if new carbohydrate compositions could prolong the lifetime of the gas bubbles, α-d-galactose was mixed with other carbohydrates or amphiphiles with varying log P. Acoustic attenuation vs. time (390 s) area under the curve (A(390)) and bubble half-time (t½) were used as measures of prolonged lifetime of gas bubbles. The products, to which 0.1% of a lipophilic carboxylic acid (5β-cholanic acid, behenic acid, and melissic acid) has been added, showed more than 5, 7 and 11 times enhancement of A(390), respectively, compared with the reference compound 2 (RC2) corresponding to the commercial product Levovist®. The half-time t ½ of the same compounds was prolonged more than 6 times compared with RC2. A partial least square (PLS) statistical analysis confirmed that, for additives, high log P carboxylic acids lead to the highest A(390). The present results bear a promise of products with a more persistent in vivo ultrasound contrast effect than the commercially available agents.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
10,12-Tricosadiynoic acid, ≥98.0% (GC)