Merck
TAK1 regulates autophagic cell death by suppressing the phosphorylation of p70 S6 kinase 1.
Scientific reports (2013-03-28)
Ju Hyun Shin, Sang-Hyun Min, Seong-Jin Kim, Young-Il Kim, Junsoo Park, Heung Kyu Lee, Ook Joon Yoo
ABSTRACT
There is growing interest in identifying regulators of autophagy. The molecular mechanism underlying transforming growth factor-β activated kinase 1 (TAK1)-induced autophagy is poorly understood. We found that TAK1 inhibits p70 S6 kinase1 (S6K1) phosphorylation by interfering interaction of raptor with S6K1, thus inducing autophagy. The factors that determine whether autophagy is cytoprotective or cytotoxic have not been fully elucidated. In Drosophila, TAK1 overexpression leads to an impaired eye phenotype despite inhibition of apoptosis, indicating that the phenotype was mainly due to autophagy. Also, TAK1 overexpression increases lactate dehydrogenase (LDH) level in mammalian cells. When treated with autophagy inhibitors, the level of TAK1-induced cytotoxicity or cell death was significantly attenuated, indicating that TAK1 induces cytotoxic autophagic cell death. This study provides the first in vitro and in vivo evidence of TAK1-induced autophagy and we believe that our findings significantly contribute to the understanding of the mechanisms underlying the induction of autophagy.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human RPS6KB1