Merck
  • Chromogranin/secretogranin proteins in murine heart: myocardial production of chromogranin A fragment catestatin (Chga(364-384)).
Chromogranin/secretogranin proteins in murine heart: myocardial production of chromogranin A fragment catestatin (Chga(364-384)).
Cell and tissue research (2010-11-06)
Nilima Biswas, Erica Curello, Daniel T O'Connor, Sushil K Mahata
ABSTRACT
In the heart, the secretory granules containing the atrial natriuretic peptides (ANP) and B-type myocardial natriuretic peptide (BNP) provide the basis for the endocrine function of this organ. We sought to determine whether atrial and myocardial secretory granules contain chromogranin/secretogranin proteins including chromogranin A (CHGA/Chga), chromogranin B (CHGB/Chgb) and secretogranin II (SCG2/Scg2). Deconvolution microscopy on immunolabeled proteins revealed the presence of Chga, Chgb, and Scg2 in murine cardiac secretory granules. The presence of low plasma catestatin (CST: mChga(364-384)) in older mice indicates diminished processing of Chga to CST with advancement of age, which is comparable to that found in humans. We have previously shown that CST (hCHGA(352-372)) exerts potent cardio-suppressive effects on frog and rat heart, but the source of CST for such action has remained elusive. In the present study, we found CST-related peptides in cardiomyocytes and in heart, which establishes an autocrine/paracrine function of CST in cardiac tissue. We conclude that cardiac secretory granules contain Chga, Chgb and Scg2 and that Chga is processed to CST in murine heart.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human CHGB