Merck
  • Wilms' tumor gene 1 (WT1) silencing inhibits proliferation of malignant peripheral nerve sheath tumor sNF96.2 cell line.
Wilms' tumor gene 1 (WT1) silencing inhibits proliferation of malignant peripheral nerve sheath tumor sNF96.2 cell line.
PloS one (2014-12-05)
Rosalba Parenti, Venera Cardile, Adriana Carol Eleonora Graziano, Carmela Parenti, Assunta Venuti, Maria Paola Bertuccio, Debora Lo Furno, Gaetano Magro
ABSTRACT
Wilms' tumor gene 1 (WT1) plays complex roles in tumorigenesis, acting as tumor suppressor gene or an oncogene depending on the cellular context. WT1 expression has been variably reported in both benign and malignant peripheral nerve sheath tumors (MPNSTs) by means of immunohistochemistry. The aim of the present study was to characterize its potential pathogenetic role in these relatively uncommon malignant tumors. Firstly, immunohistochemical analyses in MPNST sNF96.2 cell line showed strong WT1 staining in nuclear and perinuclear areas of neoplastic cells. Thus, we investigated the effects of silencing WT1 by RNA interference. Through Western Blot analysis and proliferation assay we found that WT1 knockdown leads to the reduction of cell growth in a time- and dose-dependent manner. siWT1 inhibited proliferation of sNF96.2 cell lines likely by influencing cell cycle progression through a decrease in the protein levels of cyclin D1 and inhibition of Akt phosphorylation compared to the control cells. These results indicate that WT1 knockdown attenuates the biological behavior of MPNST cells by decreasing Akt activity, demonstrating that WT1 is involved in the development and progression of MPNSTs. Thus, WT1 is suggested to serve as a potential therapeutic target for MPNSTs.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-actina, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-WT1 Antibody, clone 6F-H2, clone 6F-H2, Upstate®, from mouse