Merck
  • Synthesis of axially chiral oxazoline-carbene ligands with an N-naphthyl framework and a study of their coordination with AuCl·SMe(2).
Synthesis of axially chiral oxazoline-carbene ligands with an N-naphthyl framework and a study of their coordination with AuCl·SMe(2).
Beilstein journal of organic chemistry (2012-09-28)
Feijun Wang, Shengke Li, Mingliang Qu, Mei-Xin Zhao, Lian-Jun Liu, Min Shi
ABSTRACT
Axially chiral oxazoline-carbene ligands with an N-naphthyl framework were successfully prepared, and their coordination behavior with AuCl·SMe(2) was also investigated, affording the corresponding Au(I) complexes in moderate to high yields.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Methyl 1-hydroxy-2-naphthoate, 98%