Merck
Reconstituted B cell receptor signaling reveals carbohydrate-dependent mode of activation.
Scientific reports (2016-11-01)
Rina F Villar, Jinal Patel, Grant C Weaver, Masaru Kanekiyo, Adam K Wheatley, Hadi M Yassine, Catherine E Costello, Kevin B Chandler, Patrick M McTamney, Gary J Nabel, Adrian B McDermott, John R Mascola, Steven A Carr, Daniel Lingwood
ABSTRACT
Activation of immune cells (but not B cells) with lectins is widely known. We used the structurally defined interaction between influenza hemagglutinin (HA) and its cell surface receptor sialic acid (SA) to identify a B cell receptor (BCR) activation modality that proceeded through non-cognate interactions with antigen. Using a new approach to reconstitute antigen-receptor interactions in a human reporter B cell line, we found that sequence-defined BCRs from the human germline repertoire could be triggered by both complementarity to influenza HA and a separate mode of signaling that relied on multivalent ligation of BCR sialyl-oligosaccharide. The latter suggested a new mechanism for priming naïve B cell responses and manifested as the induction of SA-dependent pan-activation by peripheral blood B cells. BCR crosslinking in the absence of complementarity is a superantigen effect induced by some microbial products to subvert production of antigen-specific immune responses. B cell superantigen activity through affinity for BCR carbohydrate is discussed.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anticorpo anti-fosfotirosina, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anti-Human IgM (μ-chain specific)−Agarose antibody produced in goat, affinity isolated antibody, PBS suspension