Merck
Epstein-Barr virus maintains lymphomas via its miRNAs.
Oncogene (2013-03-19)
D T Vereide, E Seto, Y-F Chiu, M Hayes, T Tagawa, A Grundhoff, W Hammerschmidt, B Sugden
ABSTRACT
Epstein-Barr virus (EBV) has evolved exquisite controls over its host cells, human B lymphocytes, not only directing these cells during latency to proliferate and thereby expand the pool of infected cells, but also to survive and thereby persist for the lifetime of the infected individual. Although these activities ensure the virus is successful, they also make the virus oncogenic, particularly when infected people are immunosuppressed. Here we show, strikingly, that one set of EBV's microRNAs (miRNAs) both sustain Burkitt's lymphoma (BL) cells in the absence of other viral oncogenes and promote the transformation of primary B lymphocytes. BL cells were engineered to lose EBV and found to die by apoptosis and could be rescued by constitutively expressing viral miRNAs in them. Two of these EBV miRNAs were found to target caspase 3 to inhibit apoptosis at physiological concentrations.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Monoclonal Anti-IPO7, (C-terminal) antibody produced in mouse, clone 4G6, purified immunoglobulin, buffered aqueous solution