Merck
  • Role of PCSK9 and IDOL in the pathogenesis of acquired LDL receptor deficiency and hypercholesterolemia in nephrotic syndrome.
Role of PCSK9 and IDOL in the pathogenesis of acquired LDL receptor deficiency and hypercholesterolemia in nephrotic syndrome.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association (2013-10-30)
Shuman Liu, Nosratola D Vaziri
ABSTRACT
Nephrotic syndrome (NS) leads to elevation of serum total and LDL cholesterol. This is largely due to impaired LDL clearance, which is caused by hepatic LDL receptor (LDLR) deficiency despite normal LDLR mRNA expression, pointing to a post-transcriptional process. The mechanism(s) by which NS causes LDLR deficiency is not known. By promoting degradation of LDLR, Proprotein Convertase Subtilisin/Kexin type 9 (PCSK9) and inducible degrader of the LDL receptor (IDOL) play a major role in post-translational regulation of LDLR. We, therefore, tested the hypothesis that LDLR deficiency despite its normal gene expression in NS may be due to upregulation of hepatic PCSK9 and IDOL. LDLR, IDOL and PCSK9 expressions and nuclear translocation of liver X receptor (LXR) that regulates IDOL expression were determined in the liver of rats with puromycin-induced NS and control (CTL) rats. Compared with the CTLs, the NS rats showed marked elevation of serum total and LDL cholesterol and a significant reduction in hepatic LDLR protein expression. This was accompanied by marked upregulation of hepatic PCSK9 and IDOL expressions and heightened LXR activation. LDLR deficiency, hypercholesterolemia and elevated plasma LDL in NS are associated with upregulation of PCSK9 and IDOL. Interventions targeting these pathways may be effective in the management of hypercholesterolemia and the associated cardiovascular and other complications of NS.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-MYLIP antibody produced in rabbit, affinity isolated antibody