- Short-Term Ketamine Treatment Decreases Oxidative Stress Without Influencing TRPM2 and TRPV1 Channel Gating in the Hippocampus and Dorsal Root Ganglion of Rats.
Short-Term Ketamine Treatment Decreases Oxidative Stress Without Influencing TRPM2 and TRPV1 Channel Gating in the Hippocampus and Dorsal Root Ganglion of Rats.
Cellular and molecular neurobiology (2016-03-05)
Arif Demirdaş, Mustafa Nazıroğlu, Ishak Suat Övey
PMID26935063
ABSTRACT
Calcium ions (Ca
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto