Merck
Targeting IL-6 for the treatment of rheumatoid arthritis: Phase II investigational drugs.
Expert opinion on investigational drugs (2014-04-29)
Luca Semerano, Allan Thiolat, Emeline Minichiello, Gaëlle Clavel, Natacha Bessis, Marie-Christophe Boissier
ABSTRACT
IL-6 is a key cytokine in the pathogenesis of rheumatoid arthritis (RA). The clinical efficacy of tocilizumab (TCZ), a humanized anti-IL6-receptor mAb, confirmed the value of IL-6 blockade in this disease. A number of new anti-IL-6 biologics are currently in Phase I - III of clinical development for RA. This article reviews the available results from Phase II trials of investigational anti-IL-6 agents in RA. The authors discuss the potential relevance of alternative IL-6-blocking agents, with regard to their specific molecular targets in IL-6 signaling pathways and to the main open questions in the clinical research agenda for anti-IL-6 biologics. The results of Phase II trials of new anti-IL-6 biologics show promising results in terms of efficacy. The most frequently reported adverse events were not unexpected based on previous experience with TCZ. Further evidence is needed to appraise whether the difference in molecular structure or in the specific target of new anti-IL-6 biologics might result in added therapeutic value over TCZ. New data from Phase III trials that provides a head-to-head comparison against TCZ and anti-TNF agents with or without methotrexate background treatment are expected in the future.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Graphene ink in water, inkjet printable
Sigma-Aldrich
SILuRich Anti-IL6 MAb, biotin conjugated, clone IL6-4G1, purified from hybridoma cell culture