Merck
  • Metal inhibition on the reactivity of manganese dioxide toward organic contaminant oxidation in relation to metal adsorption and ionic potential.
Metal inhibition on the reactivity of manganese dioxide toward organic contaminant oxidation in relation to metal adsorption and ionic potential.
Chemosphere (2016-12-16)
Jing Jiang, Zhuopu Wang, Yang Chen, Anfei He, Jianliang Li, G Daniel Sheng
ABSTRACT
Coexisting metal ions may significantly inhibit the oxidative reactivity of manganese oxides toward organic contaminants in metal-organic multi-pollutant waters. While the metal inhibition on the oxidation of organic contaminants by manganese oxides has previously been reported, the extent of the inhibition in relation to metal properties has not been established. Six alkali, alkaline, and transition metals, as well as two testing metals were evaluated for their abilities to inhibit the reactivity of birnessite. Regardless of the pathways of phenol and diuron oxidation (polymerization vs. breakdown), the extent of metal inhibition depended mainly on the metal itself and its concentration. The observed metal inhibition efficiency followed the order of Mn
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Sodium permanganate solution, 40 wt. % in H2O