Merck
Determination of acid dissociation constants (pK
Chemosphere (2016-11-30)
Alyson R Ribeiro, Torsten C Schmidt
ABSTRACT
Cefapirin (CEPA) and ceftiofur (CEF) are two examples of widely used veterinarian cephalosporins presenting multiple ionization centers. However, the acid dissociation constants (pK