Merck
  • MiR-211 inhibits invasion and epithelial-to-mesenchymal transition (EMT) of cervical cancer cells via targeting MUC4.
MiR-211 inhibits invasion and epithelial-to-mesenchymal transition (EMT) of cervical cancer cells via targeting MUC4.
Biochemical and biophysical research communications (2016-12-08)
Dongkui Xu, Shikai Liu, Liang Zhang, Lili Song
ABSTRACT
The dysregulated molecules and their involvement in lymph node metastases of cervical cancer are far from been fully revealed. In this study, by reviewing MUC4 expression in The Human Protein Atlas and retrieving gene microarray data in GEO dataset (No. GDS4664), we found that MUC4 upregulation is associated with lymph node metastasis in cervical cancer. Knockdown of MUC4 in Hela and SiHa cells significantly reduced their invasion and also reduced the mesenchymal properties. By performing bioinformatics analysis, we observed that miR-211 is a potential suppressor of MUC4, which has a predicted highly conserved binding site in the 3'UTR of MUC among mammals. The following assays confirmed that miR-211 can directly target the 3'UTR of MUC4 and inhibit its expression at both mRNA and protein levels. In addition, enforced miR-211 expression phenocopies the effects of MUC4 siRNA in inhibiting cervical cancer cell invasion and reversing EMT properties. Therefore, we infer that miR-211 is a novel miRNA with suppressive effect on MUC4 expression and can inhibit cervical cancer cell invasion and EMT.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human MUC4