Merck
CD24 Expression and differential resistance to chemotherapy in triple-negative breast cancer.
Oncotarget (2017-04-19)
Xinyu Deng, Sophia Apple, Hong Zhao, Jeongyoon Song, Minna Lee, William Luo, Xiancheng Wu, Debra Chung, Richard J Pietras, Helena R Chang
ABSTRACT
Breast cancer (BC) is a leading cause of cancer-related death in women. Adjuvant systemic chemotherapies are effective in reducing risks of recurrence and have contributed to reduced BC mortality. Although targeted adjuvant treatments determined by biomarkers for endocrine and HER2-directed therapies are largely successful, predicting clinical benefit from chemotherapy is more challenging. Drug resistance is a major reason for treatment failures. Efforts are ongoing to find biomarkers to select patients most likely to benefit from chemotherapy. Importantly, cell surface biomarkers CD44+/CD24- are linked to drug resistance in some reports, yet underlying mechanisms are largely unknown. This study focused on the potential role of CD24 expression in resistance to either docetaxel or doxorubicin in part by the use of triple-negative BC (TNBC) tissue microarrays. In vitro assays were also done to assess changes in CD24 expression and differential drug susceptibility after chemotherapy. Further, mouse tumor xenograft studies were done to confirm in vitro findings. Overall, the results show that patients with CD24-positive TNBC had significantly worse overall survival and disease-free survival after taxane-based treatment. Also, in vitro cell studies show that CD44+/CD24+/high cells are more resistant to docetaxel, while CD44+/CD24-/low cells are resistant to doxorubicin. Both in vitro and in vivo studies show that cells with CD24-knockdown are more sensitive to docetaxel, while CD24-overexpressing cells are more sensitive to doxorubicin. Further, mechanistic studies indicate that Bcl-2 and TGF-βR1 signaling via ATM-NDRG2 pathways regulate CD24. Hence, CD24 may be a biomarker to select chemotherapeutics and a target to overcome TNBC drug resistance.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human BCL2
Sigma-Aldrich
MISSION® esiRNA, targeting human NDRG2